Learn More
Biolegend Biotin anti-TFAM
Supplier:  Biolegend 850505
Encompass_Preferred
Biotin anti-TFAM [18G102B2E11]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-207-4602