Learn More
Medchemexpress LLC Biotin-ATRA-conjugate | 2226143-93-5 | 98.4% | C36H55N3O4S | 10MG
SDP
Catalog No. 5000261326
Encompass_Preferred
ATRA-biotin (biotin-ATRA-conjugate) is a biotin-labeled derivative of all-trans retinoic acid used as a tracer and affinity reagent to track ATRA distribution and interactions in biological samples. It supports biotin-streptavidin workflows and is supplied at high purity for research use only.
  • Biotin tag enables streptavidin-based detection.
  • Suitable for localization studies in cells and tissues.
  • Compatible with pull-down and affinity capture assays.
  • High purity for reproducible experimental results.
  • Available in multiple laboratory-friendly pack sizes.
Catalog No. 50-002-61326 Supplier Medchemexpress LLC Supplier No. HY14179310MG
ATRA-biotin (biotin-ATRA-conjugate) is a biotin-labeled derivative of all-trans retinoic acid used as a tracer and affinity reagent to track ATRA distribution and interactions in biological samples. It supports biotin-streptavidin workflows and is supplied at high purity for research use only.
  • Biotin tag enables streptavidin-based detection.
  • Suitable for localization studies in cells and tissues.
  • Compatible with pull-down and affinity capture assays.
  • High purity for reproducible experimental results.
  • Available in multiple laboratory-friendly pack sizes.