Learn More
Cayman Chemical BIotIn-azIde 10mg
SDP
Catalog No. 502951860
Encompass_Preferred
A form of biotin with a terminal azide group; is used to add biotin to other molecules that bear either an alkyne group, through click chemistry, or a phosphine group, using Staudinger ligation
Catalog No. 50-295-1860 Supplier Cayman Chemical Supplier No. 1304010 MG
A form of biotin with a terminal azide group; is used to add biotin to other molecules that bear either an alkyne group, through click chemistry, or a phosphine group, using Staudinger ligation