Learn More
Selleck Chemical LLC Biotin-azide E7114-5MG
SDP
Catalog No. 5000043704
Encompass_Preferred
Biotin-azide (N-(3-Azidopropyl)biotinamide) is a form of biotin with a terminal azide group Biotin-azide can be used to prepare various biotinylated conjugates via Click Chemistry Biotin-azide is a click chemistry reagent it contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-000-43704 Supplier Selleck Chemical LLC Supplier No. E71145MG
Biotin-azide (N-(3-Azidopropyl)biotinamide) is a form of biotin with a terminal azide group Biotin-azide can be used to prepare various biotinylated conjugates via Click Chemistry Biotin-azide is a click chemistry reagent it contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups