Learn More
Apexbio Technology LLC Biotin-azide, 5mg. Cas: 908007-17-0 MFCD: MFCD20134144. biotinyltation reagent
SDP
Supplier:  Apexbio Technology LLC A80135
Encompass_Preferred
Biotin azide reacts with the terminal alkynes via a copper-catalyzed click reaction, including biomolecules containing alkyne groups through azide-alkyne cycloaddition. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4988