Learn More
TARGETMOL CHEMICALS INC Biotin-azide 5MG
SDP
Catalog No. 502879181
Encompass_Preferred
Also available in 1 mL, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Biotin-azide (N-(3-Azidopropyl)biotinamide) is used for preparing various biotin affixes and identifying proteins. Purity 99.69%
Catalog No. 50-287-9181 Supplier TARGETMOL CHEMICALS INC Supplier No. T410465MG
Also available in 1 mL, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Biotin-azide (N-(3-Azidopropyl)biotinamide) is used for preparing various biotin affixes and identifying proteins. Purity 99.69%