Learn More
Cayman Chemical Biotin-azide, Purity: ≥98%, CAS: 908007-17-0, 5 mg
SDP
Catalog No. 502954764
Encompass_Preferred
A form of biotin with a terminal azide group; is used to add biotin to other molecules that bear either an alkyne group, through click chemistry, or a phosphine group, using Staudinger ligation
Catalog No. 50-295-4764 Supplier Cayman Chemical Supplier No. 130405 MG
A form of biotin with a terminal azide group; is used to add biotin to other molecules that bear either an alkyne group, through click chemistry, or a phosphine group, using Staudinger ligation