Learn More
Medchemexpress LLC Biotin-azide | 908007-17-0 | 99.84% | 500 MG
SDP
Catalog No. 5000339115
Encompass_Preferred
Biotin-azide is a form of biotin with a terminal azide group, designed for Click Chemistry applications. It enables the preparation of various biotinylated conjugates through copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This reagent has been used in vitro for post-crosslinking chemical functionalization to isolate transcriptional protein complexes from yeast cells.
  • Used in Click Chemistry to prepare biotinylated conjugates.
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Applied in post-crosslinking chemical functionalization to isolate transcriptional protein complexes.
Catalog No. 50-003-39115 Supplier Medchemexpress LLC Supplier No. HY129832500MG
Biotin-azide is a form of biotin with a terminal azide group, designed for Click Chemistry applications. It enables the preparation of various biotinylated conjugates through copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This reagent has been used in vitro for post-crosslinking chemical functionalization to isolate transcriptional protein complexes from yeast cells.
  • Used in Click Chemistry to prepare biotinylated conjugates.
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Applied in post-crosslinking chemical functionalization to isolate transcriptional protein complexes.