Learn More
Medchemexpress LLC Biotin-azide (N-(3-Azidopropyl)biotinamide) | 908007-17-0 | 99.8% | 1 ML
SDP
Catalog No. 5000339114
Encompass_Preferred
Biotin-azide (N-(3-Azidopropyl)biotinamide) is a highly pure click chemistry reagent. It features a terminal azide group, enabling its use in preparing various biotinylated conjugates. This versatile compound is compatible with both copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions, offering broad utility in research applications.
  • Enables diverse biotinylated conjugate preparation
  • Compatible with both copper-catalyzed and strain-promoted click chemistry
  • Offers high purity for reliable experimental results
  • Primarily used as a biochemical assay reagent in research
  • Cited in studies for applications such as isolating transcriptional protein complexes
Catalog No. 50-003-39114 Supplier Medchemexpress LLC Supplier No. HY12983210MM/1ML
Biotin-azide (N-(3-Azidopropyl)biotinamide) is a highly pure click chemistry reagent. It features a terminal azide group, enabling its use in preparing various biotinylated conjugates. This versatile compound is compatible with both copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions, offering broad utility in research applications.
  • Enables diverse biotinylated conjugate preparation
  • Compatible with both copper-catalyzed and strain-promoted click chemistry
  • Offers high purity for reliable experimental results
  • Primarily used as a biochemical assay reagent in research
  • Cited in studies for applications such as isolating transcriptional protein complexes