Learn More
Medchemexpress LLC Biotin-azide (N-(3-Azidopropyl)biotinamide) | 908007-17-0 | 99.8% | 25 MG
SDP
Catalog No. 5000341576
Encompass_Preferred
Biotin-azide (N-(3-Azidopropyl)biotinamide) is a form of biotin with a terminal azide group. It can be used to prepare various biotinylated conjugates via Click Chemistry. It is a click chemistry reagent, containing an azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • A form of biotin with a terminal azide group.
  • Can be used to prepare various biotinylated conjugates via click chemistry.
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
Catalog No. 50-003-41576 Supplier Medchemexpress LLC Supplier No. 25MGHY129832
Biotin-azide (N-(3-Azidopropyl)biotinamide) is a form of biotin with a terminal azide group. It can be used to prepare various biotinylated conjugates via Click Chemistry. It is a click chemistry reagent, containing an azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • A form of biotin with a terminal azide group.
  • Can be used to prepare various biotinylated conjugates via click chemistry.
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.