Learn More
CPC Scientific Biotin-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific AMYD026A
Encompass
Synthetic peptide that forms fibrils with have the same antigenic and structural features as those of native AD amyloid filaments.ONE-LETTER SEQUENCE: Biotin-DAEFRHDSGYEVHHQKLVFFAEDVGSNKMOLECULAR FORMULA: C155H223N43O48S1MOLECULAR WEIGHT:3488.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [109770-29-8], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: J. Kang et al., Nature, 325, 773 (1987)
Catalog No. 50-210-8680