Learn More
Medchemexpress LLC HY-131455A 25mg Medchemexpress, Biotin-C1-PEG3-C3-amido-C5-Gly-Arg-Gly-N3 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131455A25MG
Encompass_Preferred
Medchemexpress, HY-131455A 25mg Biotin-C1-PEG3-C3-amido-C5-Gly-Arg-Gly-N3 (TFA) CAS: Biotin-C1-PEG3-C3-amido-C5-Gly-Arg-Gly-N3 TFA is used for detection of modification site for N-myristoylated and GPI-anchored proteins in blood-stage P. falciparum. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1287