Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-C1-PEG3-C3-amido-C5-Gly-Arg-Gly-N3 trifluoroacetate | 00-00-0 | 98.0% | C38H66F3N13O11S | 10MG
SDP

Catalog No. 5000260631
Encompass_Preferred

Biotin-C1-PEG3-C3-amido-C5-Gly-Arg-Gly-N3 trifluoroacetate is an azide-containing biotinylated reagent for click chemistry labeling of proteins. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) to conjugate with alkyne-bearing probes, enabling detection of protein modification sites. Intended for research use.

  • Contains an azide group for CuAAC and SPAAC reactions.
  • Biotin tag enables affinity detection and signal amplification.
  • PEG3 linker provides hydrophilicity and spacer flexibility.
  • High purity (98.0%) suitable for labeling applications.
  • White to off-white solid with recommended handling precautions.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store sealed at -20°C; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-60631 Supplier Medchemexpress LLC Supplier No. HY131455A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-C1-PEG3-C3-amido-C5-Gly-Arg-Gly-N3 trifluoroacetate is an azide-containing biotinylated reagent for click chemistry labeling of proteins. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) to conjugate with alkyne-bearing probes, enabling detection of protein modification sites. Intended for research use.

  • Contains an azide group for CuAAC and SPAAC reactions.
  • Biotin tag enables affinity detection and signal amplification.
  • PEG3 linker provides hydrophilicity and spacer flexibility.
  • High purity (98.0%) suitable for labeling applications.
  • White to off-white solid with recommended handling precautions.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store sealed at -20°C; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.