Learn More
Medchemexpress LLC Biotin-C10-NHS ester | 887628-40-2 | MFCD05865045 | 99.5% | 524.67 | C25H40N4O6S | 5 MG
SDP
Catalog No. 5000223449
Encompass_Preferred
Biotin-C10-NHS ester is an alkyl/ether-based PROTAC linker bearing an N-hydroxysuccinimide (NHS) ester and a ten-carbon biotin spacer. It reacts with primary amines to install a biotin handle for affinity capture, target identification, and PROTAC probe construction. The compound is supplied as a solid with reported high purity and recommended low-temperature storage to maintain stability.
  • Alkyl/ether-based PROTAC linker suited for probe design.
  • NHS ester for efficient coupling to primary amines.
  • Ten-carbon biotin spacer for robust affinity capture.
  • Supplied as a solid with reported high purity.
  • Storage conditions preserve stability (powder -20°C).
Catalog No. 50-002-23449 Supplier Medchemexpress LLC Supplier No. HY1359205MG
Biotin-C10-NHS ester is an alkyl/ether-based PROTAC linker bearing an N-hydroxysuccinimide (NHS) ester and a ten-carbon biotin spacer. It reacts with primary amines to install a biotin handle for affinity capture, target identification, and PROTAC probe construction. The compound is supplied as a solid with reported high purity and recommended low-temperature storage to maintain stability.
  • Alkyl/ether-based PROTAC linker suited for probe design.
  • NHS ester for efficient coupling to primary amines.
  • Ten-carbon biotin spacer for robust affinity capture.
  • Supplied as a solid with reported high purity.
  • Storage conditions preserve stability (powder -20°C).