Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-C4-amide-C5-NH2 | 151294-96-1 | 97.0% | 314.45 | C14H26N4O2S | 100 MG
SDP

Catalog No. 5000202926
Encompass_Preferred

Biotin-C4-amide-C5-NH2 is a biotin-based, cleavable linker used to attach cytotoxins to antibodies in the synthesis of antibody-drug conjugates (ADCs). It is supplied as a solid with specified purity and storage instructions for research applications.

  • Cleavable ADC linker suitable for ADC construction and biotin labeling.
  • Solid, white to off-white appearance.
  • Molecular weight 314.45 and formula C14H26N4O2S.
  • Purity 97.0%.
  • Available in multiple pack sizes, including 100 MG.
  • Storage: store at 4°C protected from light; in solution keep at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-02926 Supplier Medchemexpress LLC Supplier No. HYW096148100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-C4-amide-C5-NH2 is a biotin-based, cleavable linker used to attach cytotoxins to antibodies in the synthesis of antibody-drug conjugates (ADCs). It is supplied as a solid with specified purity and storage instructions for research applications.

  • Cleavable ADC linker suitable for ADC construction and biotin labeling.
  • Solid, white to off-white appearance.
  • Molecular weight 314.45 and formula C14H26N4O2S.
  • Purity 97.0%.
  • Available in multiple pack sizes, including 100 MG.
  • Storage: store at 4°C protected from light; in solution keep at -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.