Learn More
Medchemexpress LLC Biotin-C5-azide | 1260586-88-6 | 98.9% | 255.34 g/mol | C10H17N5OS | 10 MG
SDP
Catalog No. 5000286754
Encompass_Preferred
Biotin-C5-Azide is a biotinylation reagent containing a terminal azide group for click chemistry. It is intended for research use to prepare biotinylated conjugates via copper-catalyzed or strain-promoted azide-alkyne cycloaddition.
Catalog No. 50-002-86754 Supplier Medchemexpress LLC Supplier No. HY15166710MG
Biotin-C5-Azide is a biotinylation reagent containing a terminal azide group for click chemistry. It is intended for research use to prepare biotinylated conjugates via copper-catalyzed or strain-promoted azide-alkyne cycloaddition.