Learn More
Medchemexpress LLC Biotin-C5-azide (decarboxybiotin-n3) | 1260586-88-6 | C10H17N5OS | 5 MG
SDP
Catalog No. 5000372176
Encompass_Preferred
Biotin-C5-Azide (DecarboxyBiotin-N3) is a biotin reagent utilized for preparing biotinylated conjugates. It operates as a click chemistry reagent, incorporating an azide group. This group enables its participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Biotin reagent
  • Can be used to prepare biotinylated conjugates
  • Click chemistry reagent
  • Contains an azide group for CuAAc and SPAAC reactions
Catalog No. 50-003-72176 Supplier Medchemexpress LLC Supplier No. HY1516675MG
Biotin-C5-Azide (DecarboxyBiotin-N3) is a biotin reagent utilized for preparing biotinylated conjugates. It operates as a click chemistry reagent, incorporating an azide group. This group enables its participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Biotin reagent
  • Can be used to prepare biotinylated conjugates
  • Click chemistry reagent
  • Contains an azide group for CuAAc and SPAAC reactions