Learn More
Medchemexpress LLC Biotin-C5-Azide (DecarboxyBiotin-N3) | 1260586-88-6 | C10H17N5OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15166750MG
Encompass_Preferred
Biotin-C5-Azide is a biotin reagent used to prepare biotinylated conjugates. It is a click chemistry reagent containing an azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Used to prepare biotinylated conjugates
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-72946