Learn More
Echelon Biosciences Research Labs BIOTIN CERAMIDE 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs S300B100UG
Encompass
Ceramides are lipid second messengers with an important role in apoptosis. Biotin-Ceramide can be bound to streptavidin coated surfaces for protein binding experiments and assay development. This item is non returnable
Catalog No. 50-720-774