Learn More
Cayman Chemical BIotIn-Cholesterol 10mg
SDP
Catalog No. 502918362
Encompass_Preferred
A biotinylated form of cholesterol; has been used in the synthesis of ergosterol peroxide derivatives; has been used in the synthesis of biotin-conjugated liposomes and micelles for drug delivery in vitro
Catalog No. 50-291-8362 Supplier Cayman Chemical Supplier No. 3885210 MG
A biotinylated form of cholesterol; has been used in the synthesis of ergosterol peroxide derivatives; has been used in the synthesis of biotin-conjugated liposomes and micelles for drug delivery in vitro