Learn More
Cayman Chemical Biotin-Cholesterol, Purity: ≥95%, CAS: 2454045-15-7, 5 mg
SDP
Catalog No. 502919069
Encompass_Preferred
A biotinylated form of cholesterol; has been used in the synthesis of ergosterol peroxide derivatives; has been used in the synthesis of biotin-conjugated liposomes and micelles for drug delivery in vitro
Catalog No. 50-291-9069 Supplier Cayman Chemical Supplier No. 388525 MG
A biotinylated form of cholesterol; has been used in the synthesis of ergosterol peroxide derivatives; has been used in the synthesis of biotin-conjugated liposomes and micelles for drug delivery in vitro