Learn More
Bioworld Biotin-Coated Microplates, 5 Plate(s)
Catalog No. 50197541
Encompass
Biotin Coated Plates can be used as the basis for a variety of biomolecular assay techniques requiring the immobilization of probes that are conjugated with NeutrAvidin Protein, streptavidin, avidin or other biotin-binding proteins.Specifications:Biotin group accessible for binding avidin, streptavidin or NeutrAvidin™ Biotin-Binding Protein Pre-blocked to reduce nonspecific binding 96-well plate format for the ultimate in convenience
Catalog No. 50-197-541 Supplier Bioworld Supplier No. 201400101
Biotin Coated Plates can be used as the basis for a variety of biomolecular assay techniques requiring the immobilization of probes that are conjugated with NeutrAvidin Protein, streptavidin, avidin or other biotin-binding proteins.Specifications:Biotin group accessible for binding avidin, streptavidin or NeutrAvidin™ Biotin-Binding Protein Pre-blocked to reduce nonspecific binding 96-well plate format for the ultimate in convenience