Learn More
Medchemexpress LLC Biotin-COG1410 TFA | 98.0% | 1 MG
SDP
Catalog No. 5000422790
Encompass_Preferred
Biotin-COG1410 TFA is a biotin-labeled COG1410. COG1410 is an apolipoprotein E-derived peptide and an apoptosis inhibitor. It exerts neuroprotective and anti-inflammatory effects in a murine model of traumatic brain injury (TBI) and can be used for the research of neurological diseases.
  • Decreases the production and release of NO and TNFα in BV2 microglia cells.
  • Exhibits significant improvement on vestibulomotor function and spatial learning and memory in mice.
  • Improves vestibulomotor function, decreases poststroke locomotor asymmetry, and decreases infarct volume of the ipsilateral hemisphere in rats.
Catalog No. 50-004-22790 Supplier Medchemexpress LLC Supplier No. HYP2136F1MG
Biotin-COG1410 TFA is a biotin-labeled COG1410. COG1410 is an apolipoprotein E-derived peptide and an apoptosis inhibitor. It exerts neuroprotective and anti-inflammatory effects in a murine model of traumatic brain injury (TBI) and can be used for the research of neurological diseases.
  • Decreases the production and release of NO and TNFα in BV2 microglia cells.
  • Exhibits significant improvement on vestibulomotor function and spatial learning and memory in mice.
  • Improves vestibulomotor function, decreases poststroke locomotor asymmetry, and decreases infarct volume of the ipsilateral hemisphere in rats.