Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-D-Glucose | ≥90.0% | 25 MG
SDP

Catalog No. 5000450614
Encompass_Preferred

Biotin-D-Glucose is a biotin-conjugated glucose derivative.

  • Molecular formula: C16H26N2O8S
  • Molecular weight: 406.45
  • Appearance: White to off-white solid
  • Purity: ≥90.0%
  • Storage conditions: -20°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Catalog No. 50-004-50614 Supplier Medchemexpress LLC Supplier No. 25MGHYB0389F
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-D-Glucose is a biotin-conjugated glucose derivative.

  • Molecular formula: C16H26N2O8S
  • Molecular weight: 406.45
  • Appearance: White to off-white solid
  • Purity: ≥90.0%
  • Storage conditions: -20°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.