Learn More
Medchemexpress LLC Biotin-d2-1 | 1217481-41-8 | 99.7% | 1 MG
SDP
Catalog No. 5000405542
Encompass_Preferred
Biotin-d2-1 is a deuterium-labeled form of Biotin, an enzyme co-factor found in trace amounts in all living cells. This compound is intended for research use only and not for sale to patients.
  • Can be utilized as a tracer in various applications.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeling has potential to influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-05542 Supplier Medchemexpress LLC Supplier No. HYB0511S1MG
Biotin-d2-1 is a deuterium-labeled form of Biotin, an enzyme co-factor found in trace amounts in all living cells. This compound is intended for research use only and not for sale to patients.
  • Can be utilized as a tracer in various applications.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeling has potential to influence pharmacokinetic and metabolic profiles of drugs.