Learn More
Medchemexpress LLC Biotin-d2 | 58-85-5 | 98.5% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0511S21MG
Encompass_Preferred
Biotin-d2 is the deuterium labeled Biotin. Biotin is an enzyme co-factor present in minute amounts in every living cell. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07559