Learn More
Medchemexpress LLC Biotin-d2 | 58-85-5 | 98.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0511S25MG
Encompass_Preferred
Biotin-d2 is the deuterium labeled Biotin, an enzyme co-factor found in minute amounts in every living cell. This product is for research use only and is not intended for patient use.
  • This compound can be utilized as a tracer.
  • It serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • The incorporation of stable heavy isotopes like hydrogen and carbon into drug molecules, as seen with deuterium, is primarily for quantitation during drug development and has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07562