Learn More
Medchemexpress LLC Biotin-DADOO (Biotinyl-3,6-dioxaoctanediamine) | 138529-46-1 | 98.0% | 1 G
SDP
Catalog No. 5000409752
Encompass_Preferred
Biotin-DADOO is an amine compound that contains biotin and functions as a biotinylation reagent. It acts as an amino component, involved in the nucleophilic ring-opening reaction of epoxidized ω-double bonds during the synthesis of biotinylated inhibitor derivatives. This reagent can be conjugated with carboxymethyl cellulose derivatives and is utilized in the synthesis of biotin-estradiol conjugates for enzyme immunoassays. It also finds application in influenza A research.
  • Functions as a biotinylation reagent.
  • Participates in nucleophilic ring-opening reactions during biotinylated inhibitor synthesis.
  • Can be conjugated with carboxymethyl cellulose derivatives.
  • Involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays.
  • Used in influenza A research.
Catalog No. 50-004-09752 Supplier Medchemexpress LLC Supplier No. HYD09801G
Biotin-DADOO is an amine compound that contains biotin and functions as a biotinylation reagent. It acts as an amino component, involved in the nucleophilic ring-opening reaction of epoxidized ω-double bonds during the synthesis of biotinylated inhibitor derivatives. This reagent can be conjugated with carboxymethyl cellulose derivatives and is utilized in the synthesis of biotin-estradiol conjugates for enzyme immunoassays. It also finds application in influenza A research.
  • Functions as a biotinylation reagent.
  • Participates in nucleophilic ring-opening reactions during biotinylated inhibitor synthesis.
  • Can be conjugated with carboxymethyl cellulose derivatives.
  • Involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays.
  • Used in influenza A research.