Learn More
Medchemexpress LLC Biotin-DADOO (Biotinyl-3,6-dioxaoctanediamine) | 138529-46-1 | 98.0% | 250 MG
SDP
Catalog No. 5000414348
Encompass_Preferred
Biotin-DADOO is a biotin-containing amine compound and a biotinylation reagent. As an amino component, it participates in the nucleophilic ring-opening reaction of epoxidized ω-double bonds during the synthesis of biotinylated inhibitor derivatives. It can be conjugated with carboxymethyl cellulose derivatives and is involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays. Biotin-DADOO can be used in the research of influenza A.
  • Is a biotin-containing amine compound.
  • Functions as a biotinylation reagent.
  • Participates in nucleophilic ring-opening reactions during the synthesis of biotinylated inhibitor derivatives.
  • Can be conjugated with carboxymethyl cellulose derivatives.
  • Is involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays.
  • Can be used in the research of influenza A.
Catalog No. 50-004-14348 Supplier Medchemexpress LLC Supplier No. 250MGHYD0980
Biotin-DADOO is a biotin-containing amine compound and a biotinylation reagent. As an amino component, it participates in the nucleophilic ring-opening reaction of epoxidized ω-double bonds during the synthesis of biotinylated inhibitor derivatives. It can be conjugated with carboxymethyl cellulose derivatives and is involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays. Biotin-DADOO can be used in the research of influenza A.
  • Is a biotin-containing amine compound.
  • Functions as a biotinylation reagent.
  • Participates in nucleophilic ring-opening reactions during the synthesis of biotinylated inhibitor derivatives.
  • Can be conjugated with carboxymethyl cellulose derivatives.
  • Is involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays.
  • Can be used in the research of influenza A.