Learn More
Medchemexpress LLC Biotin-DADOO (Biotinyl-3,6-dioxaoctanediamine) | 138529-46-1 | 98.0% | 500 MG
SDP
Catalog No. 5000410115
Encompass_Preferred
Biotin-DADOO is a biotin-containing amine compound and a biotinylation reagent. It acts as an amino component in the nucleophilic ring-opening reaction of epoxidized ω-double bonds during the synthesis of biotinylated inhibitor derivatives. This product can be conjugated with carboxymethyl cellulose derivatives and is involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays. It can also be utilized in influenza A research.
  • Captures free influenza neuraminidase on streptavidin-coated polystyrene plates.
  • Forms stable self-assembled monolayers (SAMs) on planar gold surfaces.
  • Forms a stable biotin-estradiol conjugate.
Catalog No. 50-004-10115 Supplier Medchemexpress LLC Supplier No. HYD0980500MG
Biotin-DADOO is a biotin-containing amine compound and a biotinylation reagent. It acts as an amino component in the nucleophilic ring-opening reaction of epoxidized ω-double bonds during the synthesis of biotinylated inhibitor derivatives. This product can be conjugated with carboxymethyl cellulose derivatives and is involved in the synthesis of biotin-estradiol conjugates for enzyme immunoassays. It can also be utilized in influenza A research.
  • Captures free influenza neuraminidase on streptavidin-coated polystyrene plates.
  • Forms stable self-assembled monolayers (SAMs) on planar gold surfaces.
  • Forms a stable biotin-estradiol conjugate.