Learn More
Medchemexpress LLC Biotin-DADOO TFA | 194920-57-5 | 95.0% | 100 MG
SDP
Catalog No. 5000410608
Encompass_Preferred
Biotin-DADOO TFA is a biotinylating reagent that is conjugated to CMC thiosulfate. It can be utilized for the synthesis of biotin-estradiol conjugates, which are used to determine plasma estradiol concentrations through enzyme immunoassay. This product is intended for research use only and is not sold to patients.
  • Biotinylating reagent
  • Conjugated to CMC thiosulfate
  • Used to synthesize biotin-estradiol conjugates
  • For research use only
Catalog No. 50-004-10608 Supplier Medchemexpress LLC Supplier No. HYD0980A100MG
Biotin-DADOO TFA is a biotinylating reagent that is conjugated to CMC thiosulfate. It can be utilized for the synthesis of biotin-estradiol conjugates, which are used to determine plasma estradiol concentrations through enzyme immunoassay. This product is intended for research use only and is not sold to patients.
  • Biotinylating reagent
  • Conjugated to CMC thiosulfate
  • Used to synthesize biotin-estradiol conjugates
  • For research use only