Learn More
Medchemexpress LLC Biotin-DADOO TFA | 194920-57-5 | 95.0% | 250 MG
SDP
Catalog No. 5000414117
Encompass_Preferred
Biotin-DADOO TFA is a biotinylating reagent that is conjugated to CMC thiosulfate.
  • Can be utilized to synthesize biotin-estradiol conjugates.
  • Used for the determination of plasma estradiol concentrations by enzyme immunoassay.
  • Appearance: oil.
  • Color: light yellow to brown.
  • For research use only.
Catalog No. 50-004-14117 Supplier Medchemexpress LLC Supplier No. 250MGHYD0980A
Biotin-DADOO TFA is a biotinylating reagent that is conjugated to CMC thiosulfate.
  • Can be utilized to synthesize biotin-estradiol conjugates.
  • Used for the determination of plasma estradiol concentrations by enzyme immunoassay.
  • Appearance: oil.
  • Color: light yellow to brown.
  • For research use only.