Learn More
CPC Scientific Biotin-Asp-Glu-Val-Asp-CHO 1MG
SDP
Supplier:  CPC Scientific CASP058A
Encompass
This biotinylated apopain inhibitor has successfully been used for the affinity purification, on a streptavidin-agarose column, of caspase-3 (apopain); the enzyme that cleaves the poly(ADP-ribose) polymerase (PARP). This inhibitor showed an inhibitory actONE-LETTER SEQUENCE: Biotin-DEVD-CHOMOLECULAR FORMULA: C28H42N6O12S1MOLECULAR WEIGHT:686.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [178603-73-1], RESEARCH AREA: ApoptosisREFERENCES: D.W. Nicholson et al., Nature, 376, 37 (1995)
Catalog No. 50-210-9022