Learn More
Abcam Biotin-de x tran, 10MG
Catalog No. 50996137
Encompass_Preferred
MW 73400 g/mol, Purity >98%. Membrane impermeant dextran conjugate containing covalently attached biotin molecules (Vitamin H) making it useful in streptavidin/avidin assay systems. The attached biotins are commonly used haptens that can be detected by using its strong affinity for the proteins avidin (from egg white) or streptavidin. This conjugate contains approximately 12 biotin groups/dextran (based on amine assay; with approx. 1 amine/40 glucose residues). The excess amino groups are not capped, and they can be further labeled with a fluorophore, or can be fixed in place with formaldehyde, lysine or glutaraldehyde, allowing subsequent tissue processing, such as sectioning.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-137 Supplier Abcam Supplier No. AB27537210MG
MW 73400 g/mol, Purity >98%. Membrane impermeant dextran conjugate containing covalently attached biotin molecules (Vitamin H) making it useful in streptavidin/avidin assay systems. The attached biotins are commonly used haptens that can be detected by using its strong affinity for the proteins avidin (from egg white) or streptavidin. This conjugate contains approximately 12 biotin groups/dextran (based on amine assay; with approx. 1 amine/40 glucose residues). The excess amino groups are not capped, and they can be further labeled with a fluorophore, or can be fixed in place with formaldehyde, lysine or glutaraldehyde, allowing subsequent tissue processing, such as sectioning.
The product is subject to the following: Abcam Restricted Use Statement