Learn More
Medchemexpress LLC Biotin-EDA | 111790-37-5 | 97.5% | 1 G
SDP
Catalog No. 5000341005
Encompass_Preferred
Biotin-EDA is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This compound has a molecular weight of 286.39 and appears as a solid, white to off-white substance.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Solid, white to off-white appearance
  • For research use only
Catalog No. 50-003-41005 Supplier Medchemexpress LLC Supplier No. HY1308931G
Biotin-EDA is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This compound has a molecular weight of 286.39 and appears as a solid, white to off-white substance.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Solid, white to off-white appearance
  • For research use only