Learn More
Medchemexpress LLC Biotin-EDA | 111790-37-5 | 97.5% | 100 MG
SDP
Catalog No. 5000340011
Encompass_Preferred
Biotin-EDA is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs, which consist of two different ligands connected by a linker. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs leverage the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins
Catalog No. 50-003-40011 Supplier Medchemexpress LLC Supplier No. HY130893100MG
Biotin-EDA is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs, which consist of two different ligands connected by a linker. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs leverage the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins