Learn More
Medchemexpress LLC Biotin-EDA | 111790-37-5 | 97.5% | 5 G
SDP
Catalog No. 5000340575
Encompass_Preferred
Biotin-EDA is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs themselves consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, leveraging the intracellular ubiquitin-proteasome system to degrade target proteins selectively.
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Leverages the intracellular ubiquitin-proteoteasome system to degrade target proteins selectively
  • Alkyl-chain based
Catalog No. 50-003-40575 Supplier Medchemexpress LLC Supplier No. HY1308935G
Biotin-EDA is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs themselves consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, leveraging the intracellular ubiquitin-proteasome system to degrade target proteins selectively.
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Leverages the intracellular ubiquitin-proteoteasome system to degrade target proteins selectively
  • Alkyl-chain based