Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-EDA | 111790-37-5 | 97.5% | 50 MG
SDP

Catalog No. 5000340307
Encompass_Preferred

Biotin-EDA is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. This product is for research use only and is not sold to patients.

  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Solid, white to off-white appearance
  • Soluble in DMSO (requires ultrasonic)
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-003-40307 Supplier Medchemexpress LLC Supplier No. HY13089350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-EDA is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. This product is for research use only and is not sold to patients.

  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Solid, white to off-white appearance
  • Soluble in DMSO (requires ultrasonic)
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.