Learn More
Biomatik Corporation Biotin Elisa Kit
Supplier:  Biomatik Corporation EKC31000
Encompass_Preferred
Biotin Elisa Kit for detecting Biotin in serum, plasma from human, mouse and other species. Detection Range: 12.5-200pg/ml. Sensitivity: 4.986pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9203