Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-Gastrin Releasing Peptide, human TFA | 3085.67 | 10 MG
SDP

Catalog No. 5000399359
Encompass_Preferred

Biotin-Gastrin Releasing Peptide, human TFA is a biotinylated gastrin releasing peptide (GRP). Gastrin-releasing peptide (GRP) is a neuropeptide with growth-stimulatory and tumorigenic properties.

  • Molecular weight: 3085.67
  • Chemical formula: C140H218N40O33S3.xC2HF3O2
  • Appearance: Solid
  • Soluble in DMSO at 100 mg/mL
  • Shipping: Room temperature
  • Storage (powder): -80°C for 2 years, -20°C for 1 year
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-99359 Supplier Medchemexpress LLC Supplier No. HYP3576A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-Gastrin Releasing Peptide, human TFA is a biotinylated gastrin releasing peptide (GRP). Gastrin-releasing peptide (GRP) is a neuropeptide with growth-stimulatory and tumorigenic properties.

  • Molecular weight: 3085.67
  • Chemical formula: C140H218N40O33S3.xC2HF3O2
  • Appearance: Solid
  • Soluble in DMSO at 100 mg/mL
  • Shipping: Room temperature
  • Storage (powder): -80°C for 2 years, -20°C for 1 year
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.