Learn More
Medchemexpress LLC Biotin-Gastrin Releasing Peptide, human TFA | 3085.67 | 25 MG
SDP
Catalog No. 5000397293
Encompass_Preferred
Biotin-Gastrin Releasing Peptide, human TFA is a biotinylated gastrin releasing peptide (GRP). Gastrin-releasing peptide (GRP) is a neuropeptide known for its growth-stimulatory and tumorigenic properties.
  • Biotinylated gastrin releasing peptide (GRP)
  • Neuropeptide with growth-stimulatory properties
  • Relevant for cancer and neurological disease research
  • Related to GPCR/G protein and Bombesin Receptor studies
Catalog No. 50-003-97293 Supplier Medchemexpress LLC Supplier No. 25MGHYP3576A
Biotin-Gastrin Releasing Peptide, human TFA is a biotinylated gastrin releasing peptide (GRP). Gastrin-releasing peptide (GRP) is a neuropeptide known for its growth-stimulatory and tumorigenic properties.
  • Biotinylated gastrin releasing peptide (GRP)
  • Neuropeptide with growth-stimulatory properties
  • Relevant for cancer and neurological disease research
  • Related to GPCR/G protein and Bombesin Receptor studies