Learn More
Biolegend Biotin Goat anti-hamster (Armenian) IgG
Supplier:  Biolegend 405501
Encompass_Preferred
Biotin Goat anti-hamster (Armenian) IgG [Poly4055]; Isotype: Goat Polyclonal IgG; Reactivity: Armenian Hamster; Apps: FC, ELISA, ICFC, IHC, ICC, WB; Size: 500 ug
Catalog No. 50-169-112