Learn More
Apexbio Technology LLC Biotin Goat Anti-Mouse IgG (H+L) Antibody 2X1ml
SDP
Catalog No. 5000097063
Encompass_Preferred
Biotin Goat Anti-Mouse IgG HplusL Antibody is an affinity-purified biotin-conjugated secondary antibody targeting mouse IgG heavy and light chain regions It is designed to bind specifically to primary antibodies of mouse origin enabling signal amplification in immunological assays via biotin-avidin/streptavidin interactions Biotin Goat Anti-Mouse IgG HplusL Antibody facilitates detection in experimental models utilizing techniques such as Western blot Immunohistochemistry Immunofluorescence Flow Cytometry and ELISA by recognizing mouse IgG Based on these characteristics Biotin Goat Anti-Mouse IgG HplusL Antibody is applicable in protein detection localization quantification and cellular analysis in biomedical research
Catalog No. 50-000-97063 Supplier Apexbio Technology LLC Supplier No. K12172
Biotin Goat Anti-Mouse IgG HplusL Antibody is an affinity-purified biotin-conjugated secondary antibody targeting mouse IgG heavy and light chain regions It is designed to bind specifically to primary antibodies of mouse origin enabling signal amplification in immunological assays via biotin-avidin/streptavidin interactions Biotin Goat Anti-Mouse IgG HplusL Antibody facilitates detection in experimental models utilizing techniques such as Western blot Immunohistochemistry Immunofluorescence Flow Cytometry and ELISA by recognizing mouse IgG Based on these characteristics Biotin Goat Anti-Mouse IgG HplusL Antibody is applicable in protein detection localization quantification and cellular analysis in biomedical research