Learn More
REVMAB BIOSCIENCES USA INC Biotin IgG Fc Goat Anti-Rabbit Monoclonal Antibody [RMG02]
Supplier:  REVMAB BIOSCIENCES USA INC 41101502
Encompass_Preferred
Biotin Anti-Rabbit IgG Fc goat monoclonal antibody, Clone RMG02 (50UG) ; WB (nonreduced), IP, ICC, IHC, FC, ELISA validated; Product info: www.revmab.com
Catalog No. NC1729062