Learn More
Cayman Chemical Biotin-H10 (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502919311
Encompass_Preferred
A biotinylated form of H10; has been used in ELISA and co-immunoprecipitation assays to assess AGR2 levels in cancer cells
Catalog No. 50-291-9311 Supplier Cayman Chemical Supplier No. 370055 MG
A biotinylated form of H10; has been used in ELISA and co-immunoprecipitation assays to assess AGR2 levels in cancer cells