Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD38 Protein, Avi Tag (Avitag™);HEK293;25UG;CD8,CD38,T10,cADPr hydrolase 1
Catalog No. 501253798
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD38 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human CD38, His Tag (CD8-H82E7) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # NP_001766.2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 32.5 kDa. The protein migrates as 45-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD38 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD38, His Tag (Cat. No. CD8-H82E7) with a linear range of 0.08-1.28 μg/mL (QC tested).
Catalog No. 50-125-3798 Supplier ACROBiosystems Supplier No. CD8H82E725UG
Recombinant Protein;Biotinylated Human CD38 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human CD38, His Tag (CD8-H82E7) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # NP_001766.2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 32.5 kDa. The protein migrates as 45-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD38 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD38, His Tag (Cat. No. CD8-H82E7) with a linear range of 0.08-1.28 μg/mL (QC tested).