Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD47 Protein, Avi Tag (Avitag™);HEK293;25UG;CD7,CD47,MER6,IAP,OA3
Supplier:  ACROBiosystems CD7H82E925UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD47 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human CD47, His Tag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 16.9 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD47 MAb, Human IgG4 at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD47, His Tag (Cat. No. CD7-H82E9) with a linear range of 0.07-0.84 ng/mL (QC tested).;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD47, His Tag (Cat. No. CD7-H82E9) with a linear range of 0.8-13.1 ng/mL (Routinely tested).
Catalog No. 50-125-3823