Learn More
Medchemexpress LLC Biotin-hexanamide-(L-Thyroxine) | 2278192-78-0 | 98.6% | 1116.32 | 25 MG
SDP
Catalog No. 5000384372
Encompass_Preferred
Biotin-hexanamide-(L-Thyroxine) is a biotinylated L-Thyroxine, a synthetic hormone used for the research of hypothyroidism.
  • Biotinylated L-Thyroxine derivative.
  • Synthetic hormone for hypothyroidism research.
  • Targeted to thyroid hormone receptor.
  • Stable under recommended storage conditions.
  • Soluble in DMSO and DMF for in vitro studies.
Catalog No. 50-003-84372 Supplier Medchemexpress LLC Supplier No. 25MGHY18341F1
Biotin-hexanamide-(L-Thyroxine) is a biotinylated L-Thyroxine, a synthetic hormone used for the research of hypothyroidism.
  • Biotinylated L-Thyroxine derivative.
  • Synthetic hormone for hypothyroidism research.
  • Targeted to thyroid hormone receptor.
  • Stable under recommended storage conditions.
  • Soluble in DMSO and DMF for in vitro studies.