Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-hexanamide-(L-Thyroxine) | 2278192-78-0 | 98.6% | 1116.32 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY18341F15MG

Encompass_Preferred

Biotin-hexanamide-(L-Thyroxine) is biotinylated L-Thyroxine (HY-18341). L-Thyroxine (Levothyroxine; T4) is a synthetic hormone used for the research of hypothyroidism.

  • Biotinylated L-Thyroxine
  • Synthetic hormone for hypothyroidism research
  • Appears as a white to off-white solid
  • Store at 4°C, protected from light
  • Soluble in DMSO and DMF

Catalog No. 50-003-87563


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.