Learn More
Medchemexpress LLC Biotin-hexanamide (L-thyroxine) | 2278192-78-0 | MFCD27992063 | 98.6% | 1116.32 g·mol-1 | C31H36I4N4O7S | 10 MG
SDP
Catalog No. 5000295265
Encompass_Preferred
Biotin-hexanamide-(L-Thyroxine) is a biotinylated derivative of L-thyroxine supplied for research use. It is intended as a biochemical probe in studies of thyroid hormone biology and in assays where a biotin tag facilitates affinity capture, detection, or immobilization.
Catalog No. 50-002-95265 Supplier Medchemexpress LLC Supplier No. HY18341F110MG
Biotin-hexanamide-(L-Thyroxine) is a biotinylated derivative of L-thyroxine supplied for research use. It is intended as a biochemical probe in studies of thyroid hormone biology and in assays where a biotin tag facilitates affinity capture, detection, or immobilization.